missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSF™ Anti-Plastic Safety Shoes with Polyurethane Soles
£106.00 - £109.00
Specifications
Certifications/Compliance | EN20345 |
---|---|
Color | Black |
Footbed Material | Polyurethane |
Material | Anti-Plastic |
Sole Features | Made up of Polyurethane, water resistant |
Specifications
EN20345 | |
Polyurethane | |
Made up of Polyurethane, water resistant | |
Steel | |
Yes |
Black | |
Anti-Plastic | |
Polyurethane | |
Heat, Water-resistant | |
Safety Shoe |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title