missing translation for 'onlineSavingsMsg'
Learn More

anti-PLCG2(Phospho Y1217), PLCG2 Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DP0295.1Set

Additional Details : Weight : 0.00010kg

Product Code. 16141654

  • £581.00 / 1 set
Estimated Shipment: 06-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Phospho-PLCG2 Y1217 rabbit polyclonal antibody (20μL) In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol); PLCG2 mouse purified polyclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

Description

Description

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the PLCG2(Phospho Y1217) protein, and the other against the PLCG2 protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Specifications

PLCG2
Monoclonal/Polyclonal
PLCG2
Mouse, Rabbit
RUO
5336
Human
PLCG2(Phospho Y1217) & PLCG2 Protein Phosphorylation Antibody Pair
In situ PLA
Unconjugated
PLCG2
1 Set
Primary
Antibody pair set content: 1. Phospho-PLCG2 Y1217 rabbit polyclonal antibody (20 ul) In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (0.02% sodium azide, 50% glycerol) 2. PLCG2 mouse purified polyclonal antibody (40 ug) In 1x PBS, pH 7.2 *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers