missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-PRRX2 Ab-2 Matched Antibody Pair, Abnova™
Brand: Abnova H00051450-AP21.1Set
Additional Details : Weight : 0.00010kg
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-PRRX2 (100μg); Detection antibody: mouse purified polyclonal anti-PRRX2 (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PRRX2
Host Species: Rabbit, Mouse
Species Reactivity: Hu
ELISA Pair
Specifications
PRRX2 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PRRX2. | |
MGC19843/PMX2/PRX2 | |
Mouse, Rabbit | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-PRRX2 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-PRRX2 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
ELISA | |
Unconjugated | |
PRRX2 | |
PRRX2 | |
Affinity Purified | |
RUO | |
51450 | |
Human |