Learn More
PTK2, Human, IP-WB Antibody Pair (PW1), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00005747-PW1.1Set
Additional Details : Weight : 0.00010kg
Product Code. 16078508
  • £660.00 / 1 set
Estimated Shipment: 02-08-2024
to see stock.
Includes: Antibody pair for IP: mouse monoclonal anti-PTK2 (300μg), Antibody pair for WB: rabbit polyclonal anti-PTK2 (50μL)
Specifications
PTK2
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
FADK/FAK/FAK1/pp125FAK
Mouse, Rabbit
RUO
5747
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
PTK2
PTK2
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-PTK2 (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-PTK2 (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.