missing translation for 'onlineSavingsMsg'
Learn More

anti-PTK2, YES1 Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DI0398.1Set

Additional Details : Weight : 0.00010kg

Product Code. 16121334

  • £529.00 / 1 set
Estimated Shipment: 06-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: PTK2 rabbit purified polyclonal antibody (20μg); YES1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

Description

Description

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the PTK2 protein, and the other against the YES1 protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Specifications

PTK2
Monoclonal/Polyclonal
PTK2
PTK2
1 Set
Primary
Antibody pair set content: 1. PTK2 rabbit purified polyclonal antibody (20 ug) 2. YES1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Purified
In situ PLA
Unconjugated
FADK/FAK/FAK1/pp125FAK
Mouse, Rabbit
RUO
5747/7525
Human
PTK2 & YES1 Protein Protein Interaction Antibody Pair
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers