Learn More
anti-PTK2B Matched Antibody Pair, Abnova™
Brand: Abnova H00002185-AP51.1Set
Additional Details : Weight : 0.00010kg
Includes: Capture antibody: mouse monoclonal anti-PTK2B, IgG2b κ (100μg); Detection antibody: rabbit purified polyclonal anti-PTK2B (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PTK2B
Host Species: Mouse, Rabbit
Isotype: IgG2b κ, IgG
Species Reactivity: Hu
ELISA Pair (Transfected Lysate)
Specifications
PTK2B | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human PTK2B. | |
CADTK/CAKB/FADK2/FAK2/FRNK/PKB/PTK/PYK2/RAFTK | |
Mouse, Rabbit | |
RUO | |
2185 | |
Human | |
Purified |
ELISA | |
Unconjugated | |
PTK2B | |
PTK2B | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-PTK2B, IgG2b Kappa (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-PTK2B (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
IgG, IgG2b κ |