missing translation for 'onlineSavingsMsg'
Learn More

anti-PTPRR, MAPK1 Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DI0610.1Set

Additional Details : Weight : 0.00010kg

Product Code. 16141504

  • £529.00 / 1 set
Estimated Shipment: 06-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: PTPRR rabbit purified polyclonal antibody (20μg); MAPK1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols.

Description

Description

Host Species: Rabbit, Mouse
Cross Reactivity: Human

In situ PLA

Specifications

Specifications

PTPRR
Monoclonal/Polyclonal
PTPRR
PTPRR
1 Set
5801/5594
Human
PTPRR & MAPK1 Protein Protein Interaction Antibody Pair
In situ PLA
Unconjugated
DKFZp781C1038/EC-PTP/FLJ34328/MGC131968/MGC148170/PCPTP1/PTP-SL/PTPBR7/PTPRQ
Mouse, Rabbit
Primary
Antibody pair set content: 1. PTPRR rabbit purified polyclonal antibody (20 ug) 2. MAPK1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers