missing translation for 'onlineSavingsMsg'
Learn More

poliovirus receptor-related 1 (herpesvirus entry mediator C), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant PVRL1.

Brand:  Abnova H00005818-A01.50uL

Product Code. 16163935

  • £196.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Sequence: VQVNDSMYGFIGTDVVLHCSFANPLPSVKITQVTWQKSTNGSKQNVAIYNPSMGVSVLAPYRERVEFLRPSFTDGTIRLSRLELEDEGVYICEFATFPTGNRESQLNLTV
Specifications

Specifications

poliovirus receptor-related 1 (herpesvirus entry mediator C)
Polyclonal
Mouse polyclonal antibody raised against a partial recombinant PVRL1.
PVRL1
CD111/CLPED1/ED4/HIgR/HVEC/MGC142031/MGC16207/OFC7/PRR/PRR1/PVRR/PVRR1/SK-12/nectin-1
Mouse
50 μL
Cell Adhesion
Primary
Human
Serum
ELISA, Western Blot
Unconjugated
50% glycerol
NM_002855
PVRL1
PVRL1 (NP_002846, 33 a.a. ∼ 142 a.a) partial recombinant protein with GST tag.
RUO
Yes
5818
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers