missing translation for 'onlineSavingsMsg'
Learn More

poliovirus receptor-related 2 (herpesvirus entry mediator B), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human PVRL2 protein.

Brand:  Abnova H00005819-D01P.100ug

Product Code. 16103945

  • £297.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This gene encodes a single-pass type I membrane glycoprotein with two Ig-like C2-type domains and an Ig-like V-type domain. This protein is one of the plasma membrane components of adherens junctions. It also serves as an entry for certain mutant strains of herpes simplex virus and pseudorabies virus, and it is involved in cell to cell spreading of these viruses. Variations in this gene have been associated with differences in the severity of multiple sclerosis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq

Sequence: MARAAALLPSRSPPTPLLWPLLLLLLLETGAQDVRVQVLPEVRGQLGGTVELPCHLLPPVPGLYISLVTWQRPDAPANHQNVAAFHPKMGPSFPSPKPGSERLSFVSAKQSTGQDTEAELQDATLALHGLTVEDEGNYTCEFATFPKGSVRGMTWLRVIAKPKNQAEAQKVTFSQDPTTVALCISKEGRPPARISWLSSLDWEAKETQVSGTLAGTVTVTSRFTLVPSGRADGVTVTCKVEHESFEEPALIPVTLSVRYPPEVSISGYDDNWYLGRTDATLSCDVRSNPEPTGYDWSTTSGTFPTSAVAQGSQLVIHAVDSLFNTTFVCTVTNAVGMGRAEQVIFVRETPRASPRDVGPLVWGAVGGTLLVLLLLAGGSLAFILLRVRRRRKSPGGAGGGASGDGGFYDPKAQVLGNGDPVFWTPVVPGPMEPDGKDEEEEEEEEKAEKGLMLPPPPALEDDMESQLDGSLISRRAVYV
Specifications

Specifications

poliovirus receptor-related 2 (herpesvirus entry mediator B)
Polyclonal
Rabbit polyclonal antibody raised against a full-length human PVRL2 protein.
PVRL2
CD112/HVEB/PRR2/PVRR2
Rabbit
Affinity Purified
RUO
5819
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG
Western Blot
Unconjugated
PBS with no preservative; pH 7.4
NM_002856.2
PVRL2
PVRL2 (NP_002847.1, 1 a.a. ∼ 479 a.a) full-length human protein.
100 μg
Primary
Human
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers