missing translation for 'onlineSavingsMsg'
Learn More

anti-PXN, PIK3R1 Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DI0467.1Set

Additional Details : Weight : 0.00010kg

Product Code. 16101394

  • £529.00 / 1 set
Estimated Shipment: 06-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: PXN rabbit purified polyclonal antibody (20μg); PIK3R1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

Description

Description

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the PXN protein, and the other against the PIK3R1 protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Specifications

PXN
Monoclonal/Polyclonal
PXN
PXN
1 Set
Primary
Antibody pair set content: 1. PXN rabbit purified polyclonal antibody (20 ug) 2. PIK3R1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Purified
In situ PLA
Unconjugated
FLJ16691
Mouse, Rabbit
RUO
5829/5295
Human
PXN & PIK3R1 Protein Protein Interaction Antibody Pair
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers