Learn More
anti-Rabbit IgG, peroxidase-linked species-specific F(ab')2 fragment (from donkey) Secondary Antibody, Cytiva
Use for protein blotting, ELISA and immunocytochemistry applications. Anti-Rabbit IgG, peroxidase-linked species-specific F(ab')2 fragment (from donkey) Secondary Antibody, Cytiva, is a secondary antibody from donkey.
Brand: Life Science products Cytiva NA9340-1ML
Code : GE
Additional Details : Weight : 0.02340kg
Description
Host Species: Donkey
Isotype: IgG
Species Reactivity: Rb
Immunogen: Rabbit IgG
Protein Blotting, ELISA, Immunocytochemistry
Specifications
IgG F(Ab')2 | |
HRP | |
Affinity Purified | |
Secondary | |
IgG |
Monoclonal | |
Donkey | |
1 mL | |
Rabbit |
Safety and Handling
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : reaction mass of: 5-chloro2-methyl-4-isothiazolin3-one ¢EC no. 247-500-7! and 2-methyl-2H-isothiazol