Learn More
anti-Rabbit IgG, peroxidase-linked species-specific whole antibody (from donkey) Secondary Antibody, Cytiva
A highly species-specific anti-Rabbit IgG. Cytiva Lifescience™ anti-Rabbit IgG, peroxidase-linked species-specific whole antibody (from donkey) Secondary Antibody is used with Amersham™ ECL Western Blotting detection reagents.
Brand: Life Science products Cytiva NA934-100UL
Code : GE
Additional Details : Weight : 0.01170kg
Description
- Host Species: Donkey
- Isotype: IgG
- Species Reactivity: Rb
- Immunogen: Rabbit IgG
- Applications: Protein Blotting, ELISA, Immunocytochemistry
Specifications
IgG | |
HRP | |
Affinity Purified | |
Secondary |
Monoclonal | |
Donkey | |
0.1 mL | |
IgG |
Safety and Handling
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 5-chloro2-methyl-4-isothiazolin3-one, 2-methyl-2H-isothiazol3-one