missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-RELB, RELA Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0617.1Set
Additional Details : Weight : 0.00010kg
Includes: RELB rabbit purified polyclonal antibody (20μg); RELA mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols.
Description
Host Species: Rabbit, Mouse
Cross Reactivity: Human
In situ PLA
Specifications
RELB | |
Monoclonal/Polyclonal | |
RELB | |
RELB | |
1 Set | |
5971/5970 | |
Human | |
RELB & RELA Protein Protein Interaction Antibody Pair |
In situ PLA | |
Unconjugated | |
I-REL/IREL | |
Mouse, Rabbit | |
Primary | |
Antibody pair set content: 1. RELB rabbit purified polyclonal antibody (20 ug) 2. RELA mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |