missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHAMM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531870
311.34 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
This item is not returnable.
View return policy
Description
Recommended positive controls: human ESC. Predicted reactivity: Human (99%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is involved in cell motility. It is expressed in breast tissue and together with other proteins, it forms a complex with BRCA1 and BRCA2, thus is potentially associated with higher risk of breast cancer. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Specifications
RHAMM | |
Polyclonal | |
Unconjugated | |
HMMR | |
AA386826; CD168; Hmmr; Hyaluronan mediated motility receptor; hyaluronan mediated motility receptor (RHAMM); hyaluronan-mediated motility receptor; hyaluronan-mediated motility receptor (RHAMM); IHABP; Intracellular hyaluronic acid-binding protein; receptor for hyaluronan-mediated motility; RHAMM | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25460, 3161 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
O75330, P97779 | |
HMMR | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 198 of Human RHAMM | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RHAMM Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction