missing translation for 'onlineSavingsMsg'
Learn More

ring finger protein 34, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human RNF34 protein.

Brand:  Abnova H00080196-B01.50uL

Product Code. 16160197

This item is not returnable. View return policy

Description

Description

The protein encoded by this gene contains a RINF finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein interacts with DNAJA3/hTid-1, which is a DnaJ protein reported to function as a modulator of apoptosis. Overexpression of this gene in Hela cells was shown to confer the resistance to TNF-alpha induced apoptosis, suggesting an anti-apoptotic function of this protein. This protein can be cleaved by caspase-3 during the induction of apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq

Sequence: MRKAGATSMWASCCGLLNEVMGTGAVRGQQSAFAGATGPFRFTPNPEFSTYPPAATEGPNIVCKACGLSFSVFRKKHVCCDCKKDFCSVCSVLQENLRRCSTCHLLQETAFQRPQLMRLKVKDLRQYLILRNIPIDTCREKEDLVDLVLCHHGLGSEDDMDTSSLNSSRSQTSSFFTRSFFSNYTAPSATMSSFQGELMDGDQTSRSGVPAQVQSEITSANTEDDDDDDDEDDDDEEENAEDRNPGLSKERVRASLSDLSSLDDVEGMSVRQLKEILARNFVNYSGCCEKWELVEKVNRLYKENEENQKSYGERLQLQDEEDDSLCRICMDAVIDCVLLECGHMVTCTKCGKRMSECPICRQYVVRAVHVFKS
Specifications

Specifications

ring finger protein 34
Polyclonal
Mouse polyclonal antibody raised against a full-length human RNF34 protein.
RNF34
FLJ21786/RFI/RIF/RIFF
Mouse
50 μL
Ubiquitin/Proteasome
Primary
Human
Serum
Immunofluorescence, Western Blot
Unconjugated
No additive
NM_194271.1
RNF34
RNF34 (NP_919247.1, 1 a.a. ∼ 373 a.a) full-length human protein.
RUO
Yes
80196
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers