Learn More
3M™ Anti-Scratch/Anti-Fog Safety Glasses Product Code.: 15563417
3M™ Anti-Scratch/Anti-Fog Safety Glasses
Product Code. 15563417
Clear
Click to view available options
Lens Tint:
Amber
Clear
Gray
Shade 5
Product Code. 15563417
Brand: 3M™ 7000032456
Quantity Price
1 £7.86 / Each
20 £7.08 / Each
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Anti-Scratch/Anti-Fog Safety Glasses has a lightweight, sleek design offering excellent coverage and field of vision.
  • Very lightweight, sleek and modern design safety glasses offering excellent coverage and field of vision
  • Strong polycarbonate lens for impact resistance
  • Optical class 1 for maximum clarity
  • Inclinable lens for easy adjustment and maximum comfort
  • Soft, flexible temples for wearer comfort
  • Anti-Scratch and Anti-Fog coating for increased durability and improved vision
  • Combines well with 3M™ Aura™ 9300+ Series Disposable Respirators
  • Safety glasses designed with user comfort and convenience in mind
  • Strong polycarbonate lens for impact resistance
  • Lightweight (26g) for maximum comfort all day
  • Integrated browguard for increased protection
  • Soft inner padding on temple for increased comfort and stability when worn
  • Adjustable temple lengths in three positions for optimal comfort and fit
  • Inclinable lens for easy adjustment and maximum comfort
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved, EN 166:2001
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety glasses
Frame Color Dark Gray
Lens Material Polycarbonate
Product Content Correction
Product Title
3M™ Anti-Scratch/Anti-Fog Safety Glasses >