Learn More
SMAD3, Human, Matched Antibody Pair (AP43), Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00004088-AP43.1Set
Product Code. 16102314
  • £684.00 / 1 set
Explore more special offers
Includes: Capture antibody: mouse monoclonal anti-SMAD3 (100μg) , Detection antibody: biotinylated mouse monoclonal anti-SMAD3, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
SMAD3
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SMAD3.
DKFZp586N0721/DKFZp686J10186/HSPC193/HsT17436/JV15-2/MADH3/MGC60396
Mouse
RUO
4088
Human
Liquid
ELISA
Biotin
SMAD3
SMAD3
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-SMAD3 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-SMAD3, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.