missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ North™ Anti-Static Nitrile Finger Cots

Product Code. 15286688
Click to view available options
Size:
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Powder-free finger cots feature a low particulate count, and help to eliminate damaging static discharge in a wide variety of applications.

  • Patented nitrile formulation is comfortable with a high level of stretch
  • Low extractables result in less contamination from reactive ions
  • Helps to eliminate damaging static discharge in a wide variety of applications
  • Free of natural latex proteins that may cause allergic reactions
  • Uniform thickness with excellent tactile sensitivity
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OSHA Standard - 29 CFR 1915.157
Applications Industrial
Color White
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type ESD Finger Cot
Size Medium
Thickness (English) 3 mil
For Use With (Application) Manufacturing Processes
Glove Cuff Style Rolled
Length (English) 2.8 in.
Length (Metric) 7 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'