missing translation for 'onlineSavingsMsg'
Learn More
Honeywell™ North™ Anti-Static Nitrile Finger Cots Product Code.: 15296688
Add to basket

Honeywell™ North™ Anti-Static Nitrile Finger Cots

Product Code. 15296688
Large
Click to view available options
Size:
Medium
Large

Product Code. 15296688

Brand: Honeywell™ North™ 135LNR/L

Add to basket
Add to basket

Powder-free finger cots feature a low particulate count, and help to eliminate damaging static discharge in a wide variety of applications.

  • Patented nitrile formulation is comfortable with a high level of stretch
  • Low extractables result in less contamination from reactive ions
  • Helps to eliminate damaging static discharge in a wide variety of applications
  • Free of natural latex proteins that may cause allergic reactions
  • Uniform thickness with excellent tactile sensitivity
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OSHA Standard - 29 CFR 1915.157
Applications Industrial
Color White
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type ESD Finger Cot
Size Large
Thickness (English) 3 mil
For Use With (Application) Manufacturing Processes
Glove Cuff Style Rolled
Length (English) 2.8 in.
Length (Metric) 7 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Honeywell™ North™ Anti-Static Nitrile Finger Cots >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.