missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Streptavidin APC, BD
Research applications include studies of staining by indirect immunofluorescence when used with biotinylated antibodies.
Brand: BD Biosciences 349024
Additional Details : Weight : 0.05710kg
Description
- Flow Cytometry
- Volume per test: 20μL
- Source: prepared chromatographically from Streptomyces avidinii
- Stabilizer: Gelatin
- Preservative: 0.1% Sodium azide
Specifications
Flow Cytometry | |
RUO | |
Liquid |
100 Tests | |
Immunology |