missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Pharmingen™ APC Streptavidin
Brand: BD 554067
Additional Details : Weight : 0.01000kg
Description
Flow Cytometry
Specifications
0.2 mg/mL | |
0.1 mg | |
Aqueous buffered solution containing less than or equal to 0.09% sodium azide | |
Flow Cytometry |
Streptavidin | |
For research use only (RUO) | |
Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze. |