missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Horizon™ V450 Streptavidin
V450 Streptavidin is a useful second-step reagent for the indirect immunofluorescent staining of cells in combination with biotinylated primary antibodies for flow cytometric analysis.
Brand: BD 560797
Additional Details : Weight : 0.01000kg
Description
- Flow Cytometry
Specifications
Streptavidin | |
V450 | |
For research use only (RUO) | |
IGHM; MU; Ig mu chain C region; AGM1; VH | |
Flow cytometry |
0.5 mg/mL | |
50 Tests | |
Aqueous buffered solution containing protein stabilizer and less than or equal to 0.09% sodium azide | |
Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze. |