missing translation for 'onlineSavingsMsg'
Learn More

TCR β chain Hamster anti-Mouse, FITC, Clone: H57-597, BD

Hamster Monoclonal Antibody

Brand:  BD Biosciences 553171

 View more versions of this product

Product Code. 15817058

  • £458.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The H57-597 antibody reacts with a common epitope of the β; chain of the T-cell Receptor (TCR) complex on α;β; TCR-expressing thymocytes and peripheral T lymphocytes and NK1.1+ thymocytes and NK-T cells of all mouse strains tested. It does not react with γδ TCR-bearing T cells. In the fetal and adult thymus, the TCR β; -chain may form homodimers or pair with the pre-TCR α; -chain on the surface of immature thymocytes before expression of the TCR α; -chain. Plate-bound or soluble H57-597 antibody activates α;β; TCR-bearing T cells, and plate-bound mAb can induce apoptotic death.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TCR β chain
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
TCR affinity-purified from mouse T-cell hybridoma DO-11.10
0.5 mg
Immunology
Mouse
IgG2
Flow Cytometry
H57-597
FITC
Hamster
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.