Learn More
anti-TDP1 Matched Antibody Pair, Abnova
Brand:  Abnova H00055775-AP11.1Set
Additional Details : Weight : 0.00010kg
Product Code. 16065249
  • £660.00 / 1 set
Estimated Shipment: 02-08-2024
to see stock.
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-TDP1 (100μg); Mouse monoclonal anti-TDP1, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
Description
The protein encoded by this gene is involved in repairing stalled topoisomerase I-DNA complexes by catalyzing the hydrolysis of the phosphodiester bond between the tyrosine residue of topoisomerase I and the 3-prime phosphate of DNA. This protein may also remove glycolate from single-stranded DNA containing 3-prime phosphoglycolate, suggesting a role in repair of free-radical mediated DNA double-strand breaks. This gene is a member of the phospholipase D family and contains two PLD phosphodiesterase domains. Mutations in this gene are associated with the disease spinocerebellar ataxia with axonal neuropathy (SCAN1). While several transcript variants may exist for this gene, the full-length natures of only two have been described to date. These two represent the major variants of this gene and encode the same isoform.
ELISA
Specifications
Specifications
TDP1
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TDP1.
FLJ11090/MGC104252
Mouse, Rabbit
1 Set
Stem Cell Biology
55775
Human
IgG1
ELISA
Unconjugated
TDP1
TDP1
Affinity Purified
RUO
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-TDP1 (100 μg) 2. Detection antibody: mouse monoclonal anti-TDP1, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.