missing translation for 'onlineSavingsMsg'
Learn More

transforming growth factor, beta-induced, 68kDa, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant TGFBI.

Brand:  Abnova H00007045-A01.50uL

Product Code. 16186575

  • £196.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This gene encodes an RGD-containing protein that binds to type I, II and IV collagens. The RGD motif is found in many extracellular matrix proteins modulating cell adhesion and serves as a ligand recognition sequence for several integrins. This protein plays a role in cell-collagen interactions and may be involved in endochondrial bone formation in cartilage. The protein is induced by transforming growth factor-beta and acts to inhibit cell adhesion. Mutations in this gene are associated with multiple types of corneal dystrophy. [provided by RefSeq

Sequence: AHDKRGRYGTLFTMDRVLTPPMGTVMDVLKGDNRFSMLVAAIQSAGLTETLNREGVYTVFAPTNEAFRALPPRERSRLLGDAKELANILKYHIGDEILVS
Specifications

Specifications

transforming growth factor, beta-induced, 68kDa
Polyclonal
Mouse polyclonal antibody raised against a partial recombinant TGFBI.
TGFBI
BIGH3/CDB1/CDG2/CDGG1/CSD/CSD1/CSD2/CSD3/EBMD/LCD1
Mouse
50 μL
Cell Adhesion
Primary
Human
Serum
ELISA, Western Blot
Unconjugated
50% glycerol
BC000097
TGFBI
TGFBI (AAH00097.1, 481 a.a. ∼ 580 a.a) partial recombinant protein with GST tag.
RUO
Yes
7045
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.