Learn More
anti-TNFRSF1A, HSA Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0504.1Set
Additional Details : Weight : 0.00010kg
Includes: TNFRSF1A rabbit purified polyclonal antibody (20μg); HSA mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the TNFRSF1A protein, and the other against the HSA protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
TNFRSF1A | |
Monoclonal/Polyclonal | |
TNFRSF1A | |
TNFRSF1A | |
1 Set | |
Primary | |
Antibody pair set content: 1. TNFRSF1A rabbit purified polyclonal antibody (20 ug) 2. HSA mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
CD120a/FPF/MGC19588/TBP1/TNF-R/TNF-R-I/TNF-R55/TNFAR/TNFR1/TNFR55/TNFR60/p55/p55-R/p60 | |
Mouse, Rabbit | |
RUO | |
7132/7124 | |
Human | |
TNFRSF1A & HSA Protein Protein Interaction Antibody Pair |