Learn More
anti-TP53, HSPA1B Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0230.1Set
Additional Details : Weight : 0.02000kg
Includes: TP53 rabbit purified polyclonal antibody (20μg); HSPA1B mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the TP53 protein, and the other against the HSPA1B protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
TP53 | |
Monoclonal/Polyclonal | |
TP53 | |
TP53 | |
1 Set | |
Primary | |
Antibody pair set content: 1. TP53 rabbit purified polyclonal antibody (20 ug) 2. HSPA1B mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
FLJ92943/LFS1/TRP53/p53 | |
Mouse, Rabbit | |
RUO | |
7157/3304 | |
Human | |
TP53 & HSPA1B Protein Protein Interaction Antibody Pair |