missing translation for 'onlineSavingsMsg'
Learn More

anti-TP53, PPP2R1A Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DI0485.1Set

Additional Details : Weight : 0.02000kg

Product Code. 16022538

  • £529.00 / 1 set
Estimated Shipment: 06-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: TP53 rabbit purified polyclonal antibody (20μg); PPP2R1A mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols

Description

Description

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the TP53 protein, and the other against the PPP2R1A protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Specifications

TP53
Monoclonal/Polyclonal
TP53
TP53
1 Set
Primary
Antibody pair set content: 1. TP53 rabbit purified polyclonal antibody (20 ug) 2. PPP2R1A mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Purified
In situ PLA
Unconjugated
FLJ92943/LFS1/TRP53/p53
Mouse, Rabbit
RUO
7157/5518
Human
TP53 & PPP2R1A Protein Protein Interaction Antibody Pair
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers