missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TrkC Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen OST00119W
This item is not returnable.
View return policy
Description
Glycerol (1:1) may be added for additional stability. Reconstitute in 100 μl of sterile water.
a tyrosine-protein kinase receptor for neurotrophin-3.Specifications
TrkC | |
Polyclonal | |
NTRK3 | |
Neurotrophic tyrosine kinase receptor type 3, TrkC tyrosine kinase, GP145-TrkC, Trk-C, NT-3 growth factor receptor, NTRK3 | |
Sheep | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q16288, Q6VNS1, Q03351 | |
NTRK3 | |
A synthetic peptide from the extracellular domain of rat TrkC. | |
RUO | |
18213, 29613, 4916 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |