missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-cadherin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 361900
This item is not returnable.
View return policy
Description
This antibody has been successfully used in western blot, immunofluorescence, immunohistochemistry, and ELISA.
VE-cadherin Polyclonal antibody specifically detects VE-cadherin in Human, Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
VE-cadherin | |
Polyclonal | |
Unconjugated | |
CDH5 | |
7B4; 7B4 antigen; 7B4/cadherin-5; AA408225; cadherin 5; cadherin 5, type 2 (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular epithelium); cadherin-5; cadherin-5; cd144 antigen; VE cadherin; Cd144; cd144 antigen; CDH5; cell-cell adhesion protein; endothelial-specific cadherin; vascular endothelial cadherin; vascular endothelial cadherin precursor; Vec; VEcad; VE-Cad; VE-cadherin; VECD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1003, 12562, 307618 | |
-20°C | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.25 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P33151, P55284 | |
CDH5 | |
Synthetic peptide derived from a C-terminal region of the human VE-cadherin (vascular endothelial cadherin, cadherin-5, CD144) protein. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |