missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA516879
This item is not returnable.
View return policy
Description
PA5-16879 targets Wilms Tumor Protein in IHC (P) applications and shows reactivity with Human, mouse, and Rat samples. The PA5-16879 immunogen is a synthetic peptide derived from the C-terminus of human Wilm's tumor protein.
Wilm′s Tumor (WT), a sporadic and familial childhood kidney tumor, is genetically heterogeneous. Wilm′s tumor is associated with mutations of WT1, a zinc-finger transcription factor that is essential for the development of the metanephric kidney and the urogenital system. The WT1 gene is normally expressed infetal kidney and mesothelioma, and its expression has been suggested as a marker for Wilm′s tumor and mesothelioma.Specifications
WT1 | |
Polyclonal | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
P19544, P22561, P49952 | |
WT1 | |
A synthetic peptide derived from the C-terminus of human Wilm′s tumor protein | |
500 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
WT1 | |
Wilms tumor protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
22431, 24883, 7490 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction