missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Antibody Column Package: Modern Protein A (1-step Purification Protocol)

Combines affinity chromatography with desalting for the purification of human antibodies

Brand:  Cytiva 29497628

Additional Details : CAS Number : 64-17-5 Weight : 0.09500kg


Product Code. 17069213

  • £231.00 / 1 set
Estimated Shipment: 02-09-2024
to see stock.
Add to basket

Description

Description

  • Contains columns for a 1-step protocol using affinity chromatography: HiTrap™ MabSelect™ PrismA 1 × 1 mL and HiTrap™ Desalting 1 × 5 mL
  • HiTrap™ MabSelect PrismA column is prepacked with MabSelect PrismA resin for preparative purification of monoclonal and polyclonal antibodies from serum and cell culture supernatant
  • HiTrap™ Desalting column is packed with Sephadex G-25 to give quick pH neutralization of the eluted antibodies (0.1 to 1.5 mL is the recommended sample loading volume)
  • Includes instructions with guidance and tips to quickly get started
Specifications

Specifications

Antibody Column Pack
Protein A; Protein A, Alkali Stabilized
Resin Type: MabSelect
HiTrap™ MabSelect™ PrismA (1 x 1 mL), and HiTrap™ Desalting (1 x 5 mL)
Prepacked Column
Purification of Antibodies
2°C to 8°C
1 Kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Antibody package, modern Protein A

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers