missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Antibody Column Package: Modern Protein A (2-step Purification Protocol)
Combines affinity with size exclusion chromatography to achieve very high purity of human IgG
Brand: Cytiva 29497629
This item is not returnable.
View return policy
Description
- Contains columns suitable for a successful 2-step protocol of human antibodies: HiTrap™ MabSelect™ PrismA (1 × 1 mL) and HiPrep™ 16/60 Sephacryl S-300 HR
- Includes instructions with guidance and tips to quickly get started
- HiTrap™ MabSelect PrismA column is prepacked with MabSelect PrismA resin for preparative purification of monoclonal and polyclonal antibodies from serum and cell culture supernatant, providing excellent purity in one step
- A final high purity is obtained with the HiPrep 16/60 Sephacryl S-300 HR size exclusion column, which removes antibody aggregates and/or fragments
Specifications
Antibody Column Pack | |
Protein A; Protein A, Alkali Stabilized | |
Resin Type: MabSelect | |
HiTrap™ MabSelect™ PrismA (1 x 1 mL), and HiPrep™ 16/60 Sephacryl S-300 HR Columns |
Prepacked Column | |
Purification of Antibodies | |
2°C to 8°C | |
1 Kit |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Antibody package, modern Protein A (2-step protocol)
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction