Learn More
Thermo Scientific™ Antibody Purification on Protein A Column
Buffers to be used when performing antibody purification using immobilized Protein A
Brand: Thermo Scientific™ J63014.QA
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
This kit contains: 1L of 1M Tris-HCl (pH 8.0) and 500mL of 100mM Glycine (pH 3.0).Specifications
Purification Buffer | |
Ambient temperatures | |
Liquid |
Buffers to be used when performing antibody purification using immobilized Protein A. | |
1 Kit |