missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony, 99+%, powder, 0.15 mm, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Gray |
CAS | 7440-36-0 |
Melting Point | 630°C |
Boiling Point | 1635°C |
Density | 6.68 |
Assay Percent Range | 99% min. (Iodometry) |
Molecular Formula | Sb |
MDL Number | MFCD00011211 MFCD00134030 |
Quantity | 10 g |
Merck Index | 15, 683 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0437846 | Certificate of Analysis | 31/01/2025 | 192450100 |
A0437846 | Certificate of Analysis | 25/08/2023 | 192450100 |
A0416343 | Certificate of Analysis | 12/12/2022 | 192450100 |
A0432054 | Certificate of Analysis | 21/07/2021 | 192450100 |
A0432053 | Certificate of Analysis | 21/07/2021 | 192450100 |
Safety and Handling

Product Identifier
- Antimony, powder
- Warning
- Carcinogenicity Category 2
- H351-Suspected of causing cancer.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Antimony, 99+%, powder, 0.15 mm, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction