missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony ICP Standard, 1000 mg/L Sb in HNO3 5%, HF 1%, Honeywell Fluka™
Brand: Honeywell Fluka E1119-100ML
81.50 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Antimony ICP Standard | |
Standards for ICP and ICP-MS | |
<2 | |
100 mL |