Learn More
Antimony(III) acetate, 97%, Thermo Scientific Chemicals
£28.54 - £133.67
Chemical Identifiers
CAS | 6923-52-0 |
---|---|
Molecular Formula | C6H9O6Sb |
Molecular Weight (g/mol) | 298.892 |
MDL Number | MFCD00014974 |
InChI Key | JVLRYPRBKSMEBF-UHFFFAOYSA-K |
Synonym | antimony triacetate, antimony iii acetate, antimony acetate, acetic acid, antimony 3+ salt, octan antimonity czech, antimony 3+ triacetate, acetic acid, antimony 3+ salt 3:1, acetic acid, trianhydride with antimonic acid h3sbo3, acetic acid, antimony salt, octan antimonityShow More |
PubChem CID | 23354 |
IUPAC Name | antimony(3+);triacetate |
SMILES | CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sb+3] |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
11345009
|
Thermo Scientific Alfa Aesar
087657.09 |
10 g |
£28.54
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11365009
|
Thermo Scientific Alfa Aesar
087657.22 |
100 g |
£59.09
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11355009
|
Thermo Scientific Alfa Aesar
087657.36 |
500 g |
£133.67
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Antimony(III) acetate is a catalyst used in the production of synthetic fibers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
6923-52-0 | |
298.892 | |
JVLRYPRBKSMEBF-UHFFFAOYSA-K | |
23354 | |
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sb+3] |
C6H9O6Sb | |
MFCD00014974 | |
antimony triacetate, antimony iii acetate, antimony acetate, acetic acid, antimony 3+ salt, octan antimonity czech, antimony 3+ triacetate, acetic acid, antimony 3+ salt 3:1, acetic acid, trianhydride with antimonic acid h3sbo3, acetic acid, antimony salt, octan antimonityShow More | |
antimony(3+);triacetate |
Spezifikation
6923-52-0 | |
MFCD00014974 | |
Moisture sensitive | |
Decomposes in water | |
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sb+3] | |
298.892 | |
298.88 | |
Powder |
C6H9O6Sb | |
UN1549 | |
antimony triacetate, antimony iii acetate, antimony acetate, acetic acid, antimony 3+ salt, octan antimonity czech, antimony 3+ triacetate, acetic acid, antimony 3+ salt 3:1, acetic acid, trianhydride with antimonic acid h3sbo3, acetic acid, antimony salt, octan antimonity | |
JVLRYPRBKSMEBF-UHFFFAOYSA-K | |
antimony(3+);triacetate | |
23354 | |
97% | |
Antimony (III) acetate |
Sicherheit und Handhabung
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
missing translation for 'einecsNumber' : 230-043-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.