missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) bromide, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11398688
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Antimony(III) bromide is used in the production of other antimony compounds, in chemical analysis, as a mordant, and in dyeing. It can be added to polymers such as polyethylene as a fire retardant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-61-9
Molecular Formula Br3Sb
Molecular Weight (g/mol) 361.472
MDL Number MFCD00016317
InChI Key RPJGYLSSECYURW-UHFFFAOYSA-K
Synonym antimony tribromide, antimony iii bromide, antimony bromide, tribromostibine, stibine, tribromo, antimonous bromide, antimony bromide sbbr3, unii-6pm239qd86, hsdb 441, sbbr3
PubChem CID 24615
IUPAC Name tribromostibane
SMILES Br[Sb](Br)Br

Specifications

Boiling Point 280°C
Melting Point 96.6°C
Quantity 50 g
UN Number UN3260
Merck Index 14,706
Solubility Information Soluble in dilute hydrochloric acid,hydrobromic acid,carbon disulfide,alcohol,acetone,ammonia,benzene,chloroform.
Formula Weight 361.48
Percent Purity 99.5%
Refractive Index 1.74
Sensitivity Moisture sensitive
Density 4.148 g/mL
Assay (metals basis)
Chemical Name or Material Antimony(III) bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'