missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) Chloride, ≥99.0%, ACS Reagent, Honeywell Fluka™

Product Code. 15623270
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-91-9
Molecular Formula Cl3Sb
Molecular Weight (g/mol) 228.11
MDL Number MFCD00011212
InChI Key FAPDDOBMIUGHIN-UHFFFAOYSA-K
Synonym antimony trichloride, trichlorostibine, antimony chloride, antimony iii chloride, stibine, trichloro, antimontrichlorid, antimonous chloride, butter of antimony, antimony butter, caustic antimony
PubChem CID 24814
ChEBI CHEBI:74856
IUPAC Name trichlorostibane
SMILES [Cl-].[Cl-].[Cl-].[Sb+3]

Specifications

Boiling Point 283°C
Melting Point 73.4°C
Quantity 5 g
Linear Formula SbCl3
UN Number UN1733
Formula Weight 228.12g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Glass Bottle
Vapor Pressure 1mmHg (49°C)
Density 3.140g/3; Vapor Density: 7.9 (vs air)
Chemical Name or Material Antimony(Iii) Chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'