missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) ethoxide, 99.9% (metals basis)

Product Code. 11354758
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11354758

Brand: Thermo Scientific Alfa Aesar 043097.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10433-06-4 | C6H15O3Sb | 256.943 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10433-06-4
Molecular Formula C6H15O3Sb
Molecular Weight (g/mol) 256.943
MDL Number MFCD00015123
InChI Key JGOJQVLHSPGMOC-UHFFFAOYSA-N
Synonym antimony triethoxide, triethoxyantimony, acmc-20alrx, sb oet 3, antimony iii ethoxide, antimony 3+ ; ethanolate, ethanol, antimony 3+ salt 9ci, antimony 3+ tris ethoxide, antimony 3+ ion tris ethoxide
PubChem CID 66326
IUPAC Name antimony(3+);ethanolate
SMILES CC[O-].CC[O-].CC[O-].[Sb+3]

Specifications

Density 1.524 g/mL
Boiling Point 95°C (11 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3082
Sensitivity Moisture sensitive
Formula Weight 256.9
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Antimony (III) ethoxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.