Learn More
Antimony(III) fluoride, 98%, Thermo Scientific Chemicals
£23.41 - £212.06
Chemical Identifiers
CAS | 7783-56-4 |
---|---|
Molecular Formula | F3Sb |
Molecular Weight (g/mol) | 178.755 |
MDL Number | MFCD00011218 |
InChI Key | GUNJVIDCYZYFGV-UHFFFAOYSA-K |
Synonym | antimony trifluoride, trifluorostibine, antimony fluoride, antimony iii fluoride, stibine, trifluoro, trifluoroantimony, antimoine fluorure, antimonous fluoride, antimoine fluorure french, hsdb 438 |
PubChem CID | 24554 |
IUPAC Name | trifluorostibane |
SMILES | F[Sb](F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11427996
|
Thermo Scientific Alfa Aesar
A14068.18 |
50 g |
£23.41
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11437996
|
Thermo Scientific Alfa Aesar
A14068.30 |
250 g |
£67.72
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11417996
|
Thermo Scientific Alfa Aesar
A14068.0B |
1000 g |
£212.06
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Antimony(III) fluoride is used as a reagent in inorganic and organofluorine chemistry. It acts as a fluorination reagent and electroplating agent. Further, it is used in the preparation of dye, pottery, porcelains, ceramic enamels and glazes. It is utilized in the preparation of freon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAntimony(III) fluoride is used as a reagent in inorganic and organofluorine chemistry. It acts as a fluorination reagent and electroplating agent. Further, it is used in the preparation of dye, pottery, porcelains, ceramic enamels and glazes. It is utilized in the preparation of freon.
Solubility
Soluble in water, methanol and acetone. Insoluble in ammonia.
Notes
Moisture sensitive. It is incompatible with acids, perchloric acid, water and oxidizing agents.
Chemical Identifiers
7783-56-4 | |
178.755 | |
GUNJVIDCYZYFGV-UHFFFAOYSA-K | |
24554 | |
F[Sb](F)F |
F3Sb | |
MFCD00011218 | |
antimony trifluoride, trifluorostibine, antimony fluoride, antimony iii fluoride, stibine, trifluoro, trifluoroantimony, antimoine fluorure, antimonous fluoride, antimoine fluorure french, hsdb 438 | |
trifluorostibane |
Specifications
376°C | |
7783-56-4 | |
MFCD00011218 | |
14,709 | |
Soluble in water,methanol and acetone. Insoluble in ammonia. | |
F[Sb](F)F | |
178.755 | |
178.75 | |
Odorless | |
4.379 |
292°C | |
F3Sb | |
UN2923 | |
antimony trifluoride, trifluorostibine, antimony fluoride, antimony iii fluoride, stibine, trifluoro, trifluoroantimony, antimoine fluorure, antimonous fluoride, antimoine fluorure french, hsdb 438 | |
GUNJVIDCYZYFGV-UHFFFAOYSA-K | |
trifluorostibane | |
24554 | |
98% | |
Moisture sensitive | |
Antimony(III) fluoride |
Safety and Handling
GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H311+H331-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 232-009-2
missing translation for 'rtecsNumber' : CC5150000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.