missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) isopropoxide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11358028
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18770-47-3
Molecular Formula C9H21O3Sb
Molecular Weight (g/mol) 299.024
MDL Number MFCD00143623
InChI Key HYPTXUAFIRUIRD-UHFFFAOYSA-N
Synonym antimony iii isopropoxide, antimony 3+ tripropan-2-olate, triisopropoxyantimony, acmc-209epx, antimony iii propan-2-olate, antimony 3+ ; propan-2-olate, 2-propanol,antimony 3+ salt 9ci, antimony 3+ ion tris propan-2-olate, antimony 3+ tris propan-2-olate
PubChem CID 3034303
IUPAC Name antimony(3+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Sb+3]

Specifications

Color Colorless
Density 1.31 g/mL
Boiling Point 68°C (3 mmHg)
Flash Point 77°C (141°F)
Quantity 5 g
UN Number UN3082
Sensitivity Moisture sensitive
Formula Weight 298.8
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Antimony(III) isopropoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'