missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) sulfide, 98%

Product Code. 10082930 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10082930

Brand: Thermo Scientific Acros 223595000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1345-04-6 | S3Sb2 | 339.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1345-04-6
Molecular Formula S3Sb2
Molecular Weight (g/mol) 339.70
MDL Number MFCD00011217
InChI Key NVWBARWTDVQPJD-UHFFFAOYSA-N
Synonym Antimony trisulfide
IUPAC Name diantimony(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Sb+3].[Sb+3]

Specifications

Boiling Point 1150°C
Melting Point 550°C
Color Gray to Black
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 97.5% min. (Sb2S3)
Linear Formula Sb2S3
Merck Index 15, 701
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in conc. hcl and alkali hydroxides,soluble in alcohol,soluble in amminium hydrogen sulfide,insoluble in acetic acid
Formula Weight 339.67
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Antimony(III) sulfide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Antimony(III) sulfide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.