missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) sulfide, 99.999% (metals basis)

Product Code. 11381788
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11381788

Brand: Thermo Scientific Alfa Aesar 011683.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1345-04-6 | S3Sb2 | 339.70 g/mol

It is used in glitter compositions, fountain compositions, flash powder and vulcanizing rubber. It is employed in the production of ruby-colored glass and in plastics, as a fire retardant, in explosives, matches, and fireworks. It can be used to prepare a series of thioantimonous salts and Schlippe's salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1345-04-6
Molecular Formula S3Sb2
Molecular Weight (g/mol) 339.70
MDL Number MFCD00011217
InChI Key NVWBARWTDVQPJD-UHFFFAOYSA-N
IUPAC Name diantimony(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Sb+3].[Sb+3]

Specifications

Melting Point 550°C
Physical Form Powder
Quantity 2 g
UN Number UN1549
Merck Index 14,713
Formula Weight 339.68
Percent Purity 99.999%
Assay (metals basis)
Chemical Name or Material Antimony(III) sulfide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.