missing translation for 'onlineSavingsMsg'
Learn More

Antimony(III) telluride, 99.999% (metals basis)

Product Code. 11303358
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11303358

Brand: Thermo Scientific Alfa Aesar 036282.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For vacuum deposition. Activator in glass industry. Paints and Adhesives industries. Pigments, Ceramic frites. As semiconductor material, it has a large thermoelectric effect and is therefore used in solid state refrigerators. | CAS: 1327-50-0 | Sb2Te3 | 626.32 g/mol

For vacuum deposition. Activator in glass industry. Paints and Adhesives industries. Pigments, Ceramic frites. As semiconductor material, it has a large thermoelectric effect and is therefore used in solid state refrigerators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1327-50-0
Molecular Formula Sb2Te3
Molecular Weight (g/mol) 626.32
MDL Number MFCD00016320
InChI Key BPDQXJZWVBPDSN-UHFFFAOYSA-N
Synonym antimony telluride, tellanylidenestibanyl tellanyl stibanetellone, antimon iii tellurid, antimony iii telluride, antimony telluride,, antimony iii telluride, beads, antimony iii telluride, powder,-325 mesh trace metals basis
PubChem CID 6369653
IUPAC Name tellanylideneantimony;tellurium
SMILES [Sb]=[Te].[Sb]=[Te].[Te]

Specifications

Melting Point 629°C
Physical Form Lump
Quantity 50 g
UN Number UN1549
Solubility Information Insoluble in water.
Formula Weight 626.3
Percent Purity 99.999%
Packaging Plastic bottle
Specific Gravity 6.5
Density 6.5
Chemical Name or Material Antimony(III) telluride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.