missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony pieces, 5mm (0.2in) & down, 99.9999% (metals basis)
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Silver to Gray |
| Appearance | Silver Gray pieces |
| Density | 6.68 g/cm3 |
| Quantity | 10 g |
| Odor | Odorless |
| Assay Percent Range | 99.9999% (Metals basis) |
| Form | Pieces |
| Chemical Name or Material | Antimony |
RUO – Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion