missing translation for 'onlineSavingsMsg'
Learn More
Antimony pieces, 5mm (0.2in) & down, 99.9999% (metals basis), Thermo Scientific Chemicals Product Code.: 11361788

Antimony pieces, 5mm (0.2in) & down, 99.9999% (metals basis), Thermo Scientific Chemicals

Product Code. 11361788
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11361788

Brand: Thermo Scientific Alfa Aesar 011676.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-36-0
Molecular Formula Sb
Molecular Weight (g/mol) 121.76
MDL Number MFCD00011211 MFCD00134030
InChI Key WATWJIUSRGPENY-UHFFFAOYSA-N
Synonym powder, stibium metallicum, antimony, metallic, antimony, elemental, stibanylidyne, unii-9it35j3uv3, antymonowodor polish, unii-0vkz97k3ub, 0vkz97k3ub, antimonwasserstoffes germanShow MoreShow Less
PubChem CID 5354495
ChEBI CHEBI:30304
IUPAC Name antimony
SMILES [Sb]

Specifications

Color Silver to Gray
Appearance Silver Gray pieces
Assay Percent Range 99.9999% (Metals basis)
Quantity 250 g
Odor Odorless
Form Pieces
Chemical Name or Material Antimony

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX18K045 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog Number011676.30,011676.18,011676.09
Lot NumberP28I003 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011676.30,011676.18,011676.09
Lot NumberP24I018 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011676.30,011676.18,011676.09
Lot NumberP16D037 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011676.30,011676.18,011676.09
Lot NumberE07W011 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011676.30,011676.18,011676.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Antimony pieces, 5mm (0.2in) & down, 99.9999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.