missing translation for 'onlineSavingsMsg'
Learn More

Antimony, plasma standard solution, Specpure™ Sb 1000μg/mL

Product Code. 11308828
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
This item is not returnable. View return policy

Product Code. 11308828

Brand: Thermo Scientific Alfa Aesar 041682.AD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sb in H2O/tartaric acid/tr. HNO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula Sb in H2O/tartaric acid/tr. HNO3
MDL Number MFCD00134030

Specifications

Physical Form Liquid
Concentration Sb 1000 μg/mL
Quantity 50 mL
Chemical Name or Material Antimony, plasma standard solution, Specpure™

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.