missing translation for 'onlineSavingsMsg'
Learn More

Antimony potassium tartrate trihydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals

Product Code. 11410321
Click to view available options
Quantity:
25 g
100 g
500 g

Antimony potassium tartrate trihydrate is used as a antiparasitic agent, which used in the treatment of schistosomiasis and leishmaniasis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28300-74-5
Molecular Formula C8H4K2O12Sb2·3H2O
MDL Number MFCD00148863
Synonym Potassium antimonyl tartrate

Specifications

Physical Form Powder
Quantity 25 g
Assay Percent Range 99.0-103.0%
Solubility Information Soluble in water and glycerine. Insoluble in ethanol.
Formula Weight 667.87 (613.82 Anhydrous)
Grade ACS
Density 2.6
Chemical Name or Material Antimony potassium tartrate trihydrate
compliance-icons compliance-icons
Product Identifier
  • Antimony potassium tartrate trihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Potassium antimonyltartarate sesquihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.