missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1380°C |
UN Number | UN2871 |
Quantity | 250 g |
Percent Purity | 99.5% |
Mesh Size | −100 Mesh |
Odor | Odorless |
Weight | Apparent density: 2.4g/cm3 to 2.9g/cm3 |
Form | Powder |
Assay | (metals basis) |
Chemical Name or Material | Antimony powder |
Safety and Handling



Product Identifier
- Antimony powder
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Acute toxicity Category 4
- H301-Toxic if swallowed.
- H411-Toxic to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Antimony powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction